Merck
  • MITIGATION OF Cu(II) PHYTOTOXICITY TO RICE (ORYZA SATIVA) IN THE PRESENCE OF TiO₂ AND CeO₂ NANOPARTICLES COMBINED WITH HUMIC ACID.
MITIGATION OF Cu(II) PHYTOTOXICITY TO RICE (ORYZA SATIVA) IN THE PRESENCE OF TiO₂ AND CeO₂ NANOPARTICLES COMBINED WITH HUMIC ACID.
Environmental toxicology and chemistry (2015-03-17)
Yi Wang, Cheng Peng, Huaxiang Fang, Lijuan Sun, Hai Zhang, Jiabei Feng, Dechao Duan, Tingting Liu, Jiyan Shi
RESUMO
Engineered nanoparticles (NPs) and natural organic matter (NOM) in the environment may interact with background contaminants such as heavy metals and modify their bioavailability and toxicity. In the present study, the combined influences of 2 common NPs (TiO2 and CeO2 ) and humic acid (HA; as a model NOM) on Cu(II) phytotoxicity to rice were investigated by a 3-d root elongation assay performed on filter paper media. The results showed that the adsorption coefficients of bare TiO2 and CeO2 NPs (100 mg/L) toward Cu(2+) are 2.65 and 4.37, respectively, at an initial concentration of 10 mg/L, suggesting that Cu(II) could be strongly adsorbed by NPs, whereas HA-coated TiO2 and CeO2 NPs further enhanced the adsorption coefficients to 4.37 and 6.85, respectively. In addition, compared with Cu-alone treatment, the addition of bare TiO2 and CeO2 NPs (1000 mg/L) increased the length of rice root by 32.5% and 39.0%, respectively; however, the presence of HA-coated TiO2 and CeO2 NPs increased the root length by 90.2% and 100.1%, respectively, which indicated that the mitigation effect of HA-coated NPs on Cu(II) phytotoxicity was more visible than that of bare NPs. The results demonstrated that coexistence of NPs and HA significantly alleviated Cu(II) phytotoxicity as a result of a decrease in bioavailable soluble Cu(II) concentration, which contributes to an understanding of the potential behavior of NPs in the environment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Humic acid sodium salt, technical grade