Merck
  • The importance of the 6- and 7-positions of tetrahydroisoquinolines as selective antagonists for the orexin 1 receptor.
The importance of the 6- and 7-positions of tetrahydroisoquinolines as selective antagonists for the orexin 1 receptor.
Bioorganic & medicinal chemistry (2015-07-29)
David A Perrey, Ann M Decker, Jun-Xu Li, Brian P Gilmour, Brian F Thomas, Danni L Harris, Scott P Runyon, Yanan Zhang
RESUMO
Selective antagonism of the orexin 1 (OX1) receptor has been proposed as a potential mechanism for treatment of drug addiction. We have previously reported studies on the structure-activity relationships of tetrahydroisoquinoline-based antagonists. In this report, we elucidated the respective role of the 6- and 7-substitutions by preparation of a series of either 6-substituted tetrahydroisoquinolines (with no 7-substituents) or vice versa. We found that 7-substituted tetrahydroisoquinolines showed potent antagonism of OX1, indicating that the 7-position is important for OX1 antagonism (10 c, Ke = 23.7 nM). While the 6-substituted analogs were generally inactive, several 6-amino compounds bearing ester groups showed reasonable potency (26 a, Ke = 427 nM). Further, we show evidence that suggests several compounds initially displaying insurmountable antagonism at the OX1 receptor are competitive antagonists with slow dissociation rates.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,2,3,4-Tetrahydroisoquinoline, 95%