Merck
The G-protein regulator LGN modulates the activity of the NO receptor soluble guanylate cyclase.
The Biochemical journal (2012-06-14)
Swati Chauhan, Filip Jelen, Iraida Sharina, Emil Martin
RESUMO
sGC (soluble guanylate cyclase) is the main mediator of NO signalling. Biochemical and physiological studies suggest that, besides NO, in vivo regulation of sGC involves direct interaction with other proteins. Using yeast two-hybrid screening, we identified that the multidomain LGN (Leu-Gly-Asn repeat-enriched protein) interacts with both α1 and β1 sGC subunits. LGN and sGC co-localized in the cell cytoplasm, and the LGN-sGC complex was co-immunoprecipitated from cells expressing both proteins and from native tissues. Their interaction requires the N-terminal tetratricopeptide repeats of LGN, but does not require the N-terminal portions of α1 or β1 sGC subunits. Overexpression of LGN decreases the activity of cellular sGC, whereas knockdown of LGN mRNA and protein correlated with increased sGC activity. Although purified LGN interacts directly with purified sGC, the inhibitory effect in vitro is observed only after supplementation of cell lysate to the reaction. Although resting sGC and sGC activated by the stimulator BAY41-2272 have very similar LGN-IC50 values to the NO-stimulated sGC, they have a much higher Hill coefficient, suggesting co-operative binding with respect to LGN in the low-activated state of sGC. AGS3 (activator of G-protein signalling 3), the closest LGN homologue, also inhibits sGC. The interaction of sGC with these scaffolding proteins may expand the cross-talk between NO/cGMP signalling and other cellular pathways and tailor sGC function to specific tissues or signals.