Merck
  • Chemotactic peptide N-formyl-met-leu-phe activation of p38 mitogen-activated protein kinase (MAPK) and MAPK-activated protein kinase-2 in human neutrophils.
Chemotactic peptide N-formyl-met-leu-phe activation of p38 mitogen-activated protein kinase (MAPK) and MAPK-activated protein kinase-2 in human neutrophils.
The Journal of biological chemistry (1997-01-10)
E Krump, J S Sanghera, S L Pelech, W Furuya, S Grinstein
RESUMO
Activation of polymorphonuclear leukocytes (PMN) by chemotactic peptides initiates a series of functional responses that serve to eliminate pathogens. The intermediate steps that link engagement of the chemoattractant receptor to the microbicidal responses involve protein kinases that have yet to be identified. In this study we detected in human PMN the presence of p38 mitogen-activated protein kinase (MAPK), which became rapidly tyrosine phosphorylated and activated in response to the chemotactic peptide N-formyl-methionyl-leucyl-phenylalanine (fMLP). Pretreatment of PMN with wortmannin, a phosphatidylinositol 3-kinase inhibitor, or bis-indolylmaleimide, a protein kinase C antagonist, resulted in partial inhibition of p38 phosphorylation upon fMLP stimulation. Similarly, phosphorylation of p38 was only partially inhibited when the fMLP-induced cytosolic calcium transient was prevented. Stimulation of PMN by the chemoattractant also resulted in the rapid phosphorylation and activation of MAPK-activated protein kinase-2 (MAPKAPK-2), which was completely inhibited by the specific p38 inhibitor, SB203580. The physical interaction of p38 with MAPKAPK-2 was studied by coimmunoprecipitation. These two kinases were found to be associated in unstimulated PMN but dissociated upon activation of the cells by fMLP. Together these findings demonstrate the activation of p38 by chemotactic peptides in human PMN by a process involving phosphatidylinositol 3-kinase, protein kinase C, and calcium. p38, in turn, is an upstream activator of MAPKAPK-2.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)