Merck
PlexinA2 mediates osteoblast differentiation via regulation of Runx2.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2011-11-19)
Ji-Eun Oh, Hyung Joon Kim, Woo-Shin Kim, Zang Hee Lee, Hyun-Mo Ryoo, Soon Jung Hwang, Youngkyun Lee, Hong-Hee Kim
RESUMO
The imbalance between bone-resorbing osteoclasts and bone-forming osteoblasts often leads to bone destructive diseases such as osteoporosis. In contrast to the development of several antiresorptive agents for osteoporosis therapy, discovery of anabolic drugs has been difficult because of an insufficient understanding of the complex mechanism of bone formation. In a microarray analysis with mouse preosteoblast cells, we found that PlexinA2 (PlxnA2), a molecule previously known to mediate axon guidance in neural development, was upregulated by the osteogenic factor BMP2. PlxnA2-specific siRNA decreased Runx2 expression, osteoblast differentiation, and mineralization. Runx2 overexpression restored osteoblastic differentiation of PlxnA2-knockdown cells. PlxnA2 was associated with both type 1 and 2 BMP receptors, and BMP2 increased the interaction between PlxnA2 and type 1 receptors. PlxnA2 also affected Smad and Akt signaling pathways downstream of BMP2. Taken together, the results of our study reveal that PlxnA2 has a pro-osteogenic function by modulating BMP2 signaling. Therefore, PlxnA2 may be a useful target for development of bone anabolic therapeutics.