Merck
  • Cloning, expression, and mapping of PDCD9, the human homolog of Gallus gallus pro-apoptotic protein p52.
Cloning, expression, and mapping of PDCD9, the human homolog of Gallus gallus pro-apoptotic protein p52.
Cytogenetics and cell genetics (2000-01-21)
L Carim, L Sumoy, M Nadal, X Estivill, M Escarceller
RESUMO
We report the sequence, tissue distribution, and chromosome location of a novel gene, PDCD9 (programmed cell death 9). PDCD9 is the mammalian counterpart of the Gallus gallus pro-apoptotic protein p52. The human cDNA has an open reading frame of 1,314 nucleotides and was predicted to encode a protein of 438 amino acids with a calculated mass of 50 kDa. The protein sequences of chicken, mouse, and human PDCD9 are remarkably conserved. PDCD9 mRNA is expressed ubiquitously in adult tissues, displaying a stronger signal in heart, skeletal muscle, kidney, and liver. PDCD9 was mapped to chromosome 5q11.