Merck
  • Y14 positively regulates TNF-α-induced NF-κB transcriptional activity via interacting RIP1 and TRADD beyond an exon junction complex protein.
Y14 positively regulates TNF-α-induced NF-κB transcriptional activity via interacting RIP1 and TRADD beyond an exon junction complex protein.
Journal of immunology (Baltimore, Md. : 1950) (2013-07-03)
Sumihito Togi, Kaname Shiga, Ryuta Muromoto, Masaya Kato, Yuki Souma, Yuichi Sekine, Shigeyuki Kon, Kenji Oritani, Tadashi Matsuda
RESUMO
Although Y14 is known to be a component of the exon junction complex, we previously reported that Y14 regulates IL-6-induced STAT3 activation. In this study, we showed that endogenous Y14 positively regulated TNF-α-induced IL-6 expression in HeLa cells. Small interfering RNA-mediated Y14-knockdown reduced TNF-α-induced and NF-κB-mediated transcriptional activity, phosphorylation/degradation of IκBα, and nuclear localization of NF-κB/p65. As in the case of IL-6 stimuli, Y14 enhanced TNF-α-induced STAT3 phosphorylation, which is important for its nuclear retention. However, our manipulation of Y14 expression indicated that it is involved in TNF-α-induced IL-6 expression via both STAT3-dependent and -independent mechanisms. We screened signaling molecules in the TNF-α-NF-κB pathway and found that Y14 endogenously associated with receptor-interacting protein 1 (RIP1) and TNFR-associated death domain (TRADD). Overexpression of RIP1, but not TRADD, restored TNF-α-induced NF-κB activation in Y14-knockdown cells, and Y14 overexpression restored TNF-α-induced NF-κB activation in TRADD-knockdown cells, but not in RIP1-knockdown cells, indicating that Y14 lies downstream of TRADD and upstream of RIP1. Of importance, Y14 significantly enhanced the binding between RIP1 and TRADD, and this is a possible new mechanism for Y14-mediated modification of TNF-α signals. Although Y14 associates with MAGOH in the exon junction complex, Y14's actions in the TNF-α-NF-κB pathway are unlikely to require MAGOH. Therefore, Y14 positively regulates signals for TNF-α-induced IL-6 production at multiple steps beyond an exon junction complex protein.