Merck
  • Transcriptional consequences of schizophrenia candidate miR-137 manipulation in human neural progenitor cells.
Transcriptional consequences of schizophrenia candidate miR-137 manipulation in human neural progenitor cells.
Schizophrenia research (2014-02-22)
Matthew J Hill, Jacek G Donocik, Rosamond A Nuamah, Charles A Mein, Ricardo Sainz-Fuertes, Nicholas J Bray
RESUMO
MIR137, transcribed as the microRNA miR-137, is one of the leading candidate schizophrenia susceptibility genes to arise from large genome-wide association studies (GWAS) of the disorder. Recent data suggest that miR-137 modulates the expression of other schizophrenia susceptibility genes. Although bioinformatic resources are available with which to predict genes regulated by individual microRNA, there has been a lack of empirical data on genome-wide gene expression changes following miR-137 manipulation. We have therefore performed a genome-wide assessment of transcriptional changes in a human neural progenitor cell line after miR-137 over-expression and inhibition in order to elucidate molecular pathways by which genetic perturbation of miR-137 could promote susceptibility to schizophrenia. Bioinformatically-predicted miR-137 targets showed a small but highly significant down-regulation following miR-137 over-expression. Genes that were significantly down-regulated in association with miR-137 over-expression were enriched for involvement in neuronal differentiation. Differentially expressed genes that were confirmed by qPCR included others at genome-wide significant risk loci for schizophrenia (MAD1L1 and DPYD) and BDNF. These data point to molecular pathways through which genetic variation at the MIR137 locus could confer risk for schizophrenia.