Merck
  • Monocyte-derived dendritic cells from HLA-B27+ axial spondyloarthritis (SpA) patients display altered functional capacity and deregulated gene expression.
Monocyte-derived dendritic cells from HLA-B27+ axial spondyloarthritis (SpA) patients display altered functional capacity and deregulated gene expression.
Arthritis research & therapy (2014-08-22)
Alice Talpin, Félicie Costantino, Nelly Bonilla, Ariane Leboime, Franck Letourneur, Sébastien Jacques, Florent Dumont, Sonia Amraoui, Charles-Antoine Dutertre, Henri-Jean Garchon, Maxime Breban, Gilles Chiocchia
RESUMO
This study aimed to compare the functional capacity and gene expression profile of monocyte-derived dendritic cells (MD-DCs) in HLA-B27+ axial spondyloarthritis (SpA) patients and healthy controls. MD-DCs were differentiated with interleukin 4 (IL-4) and granulocyte-macrophage colony-stimulating factor (GM-CSF) for seven days, starting from purified CD14+ monocytes and stimulated with lipopolysaccharide (LPS) for six and twenty four hours. Their capacity to stimulate allogeneic CD4+ T cells from unrelated healthy donor was tested. Transcriptomic study was performed with Affymetrix HuGene 1.0 ST microarrays. Gene expression levels were compared between patients and controls using a multivariate design under a linear model (LIMMA). Real-time quantitative PCR (qRT-PCR) was performed for validation of the most striking gene expression differences. The stimulatory capacity of allogeneic CD4+ T cells by MD-DCs from SpA patients was decreased. Transcriptomic analysis revealed 81 genes differentially expressed in MD-DCs between SpA patients and controls (P <0.01 and fold-change <0.66 or >1.5). Four selected genes were validated by q ADAMTS15, CITED2, F13A1 and SELL. Expression levels of ADAMTS15 and CITED2, encoding a metallopeptidase and a transcription factor, respectively, were inversely correlated with each other (R = 0.75, P = 0.0003). Furthermore, in silico analysis identified several genes of the Wnt signaling pathway having expression co-regulated with CITED2. This study revealed altered function and gene expression pattern in MD-DCs from HLA-B27+ axial SpA. Co-expression study showed an inverse correlation between ADAMTS15 and CITED2. Moreover, the Wnt signaling pathway appeared as deregulated in SpA MD-DCs, a finding which may be connected to Th17-driven inflammatory responses.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, suitable for fluorescence, BioReagent, ≥90% (HPCE)
Sigma-Aldrich
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, ≥95% (HPLC), solid