Merck
Tumor-suppressive miR148a is silenced by CpG island hypermethylation in IDH1-mutant gliomas.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-09-17)
Sichen Li, Reshmi Chowdhury, Fei Liu, Arthur P Chou, Tie Li, Reema R Mody, Jerry J Lou, Weidong Chen, Jean Reiss, Horacio Soto, Robert Prins, Linda M Liau, Paul S Mischel, Phioanh L Nghiemphu, William H Yong, Timothy F Cloughesy, Albert Lai
RESUMO
IDH1/2-mutant gliomas harbor a distinct glioma-CpG island methylation phenotype (G-CIMP) that may promote the initiation and progression of secondary pathway gliomas by silencing tumor-suppressive genes. The potential role of tumor-suppressive microRNAs (miRNA; miR) in this process is not understood. To identify potential tumor-suppressive miRNA hypermethylated in glioma, the methylation profiles of IDH1/2(WT) gliomas (n = 11) and IDH1(MUT) glioma (n = 20) were compared by using massively parallel reduced representation bisulfite sequencing (RRBS). The methylation status of selected miRNA was validated by using targeted bisulfite sequencing (BiSEQ) in a large cohort of glioma tissue samples including 219 IDH1(WT) and 72 IDH1/2(MUT) samples. The expression of selected miRNAs was determined by using the TaqMan qPCR. Functional analyses of miR148a were conducted and target genes were identified. We identify miR148a as a novel, G-CIMP-associated miRNA whose methylation is tightly correlated with IDH1 mutation and associated with improved survival in patients with malignant glioma. We confirm that downregulation of miR148a can occur via DNA methylation. We demonstrate that IDH1 mutation provides a mechanism of miR148a methylation and downregulation, and that restoration of miR148a reduced tumorigenic properties of glioma cells, possibly by targeting DNMT1. We identify miR148a as a novel G-CIMP-associated miRNA, and provide results suggesting that miR148a restoration may have therapeutic implications.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dnmt1