Merck
  • In vitro and in vivo evaluation of 28DAP010, a novel diamidine for treatment of second-stage African sleeping sickness.
In vitro and in vivo evaluation of 28DAP010, a novel diamidine for treatment of second-stage African sleeping sickness.
Antimicrobial agents and chemotherapy (2014-05-29)
Tanja Wenzler, Sihyung Yang, Donald A Patrick, Olivier Braissant, Mohamed A Ismail, Richard R Tidwell, David W Boykin, Michael Zhuo Wang, Reto Brun
RESUMO
African sleeping sickness is a neglected tropical disease transmitted by tsetse flies. New and better drugs are still needed especially for its second stage, which is fatal if untreated. 28DAP010, a dipyridylbenzene analogue of DB829, is the second simple diamidine found to cure mice with central nervous system infections by a parenteral route of administration. 28DAP010 showed efficacy similar to that of DB829 in dose-response studies in mouse models of first- and second-stage African sleeping sickness. The in vitro time to kill, determined by microcalorimetry, and the parasite clearance time in mice were shorter for 28DAP010 than for DB829. No cross-resistance was observed between 28DAP010 and pentamidine on the tested Trypanosoma brucei gambiense isolates from melarsoprol-refractory patients. 28DAP010 is the second promising preclinical candidate among the diamidines for the treatment of second-stage African sleeping sickness.
MATERIAIS
Número do produto
Marca
Descrição do produto
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard