Merck
Genetic polymorphism of the phospholipase C epsilon 1 gene and risk of gastric cancer.
Chinese medical journal (2014-07-06)
Xinyang Liu, Xiaowei Zhang, Zhichao Wang, Jinjia Chang, Zheng Wu, Zhe Zhang, Shanshan Wang, Jin Li
RESUMO
The pathogenesis of gastric cancer (GC) involves environmental and genetic factors. Recently, two genome-wide association studies found that phospholipase C epsilon 1 (PLCE1) polymorphisms might be related to GC risk, and several studies further validated this finding. However, these studies yielded inconsistent results. A comprehensive database search was performed to identify eligible studies. Odds ratios with 95% confidence intervals were calculated to assess the strength of the association between PLCE1 rs2274223, rs753724, and rs11187842 and risk of GC. Subgroup analyses, publication bias, and sensitivity analyses were also conducted. Eleven studies (12 cohorts) were included in the meta-analysis. Based on 13 676 cases and 23 569 controls, a significant association between PLCE1 rs2274223 and GC risk was detected under various genotypic models. In the subgroup analyses, the association was significant for cardia GC, but weak for non-cardia GC. The association under the heterozygote model was detected for PLCE1 rs753724 and rs11187842 based on three studies involving 2768 cases and 3890 controls. Our findings demonstrate that the presence of the G allele at rs2274223 of the PLCE1 gene may contribute to susceptibility to GC, especially cardia GC. PLCE1 rs753724 and rs11187842 are associated with GC risk under the heterozygote model. Further well-designed large studies are warranted to validate these findings.