Merck
  • Rapid Cl⁻/HCO⁻₃exchange kinetics of AE1 in HEK293 cells and hereditary stomatocytosis red blood cells.
Rapid Cl⁻/HCO⁻₃exchange kinetics of AE1 in HEK293 cells and hereditary stomatocytosis red blood cells.
American journal of physiology. Cell physiology (2013-07-12)
Etienne Frumence, Sandrine Genetet, Pierre Ripoche, Achille Iolascon, Immacolata Andolfo, Caroline Le Van Kim, Yves Colin, Isabelle Mouro-Chanteloup, Claude Lopez
RESUMO
Anion exchanger 1 (AE1) or band 3 is a membrane protein responsible for the rapid exchange of chloride for bicarbonate across the red blood cell membrane. Nine mutations leading to single amino-acid substitutions in the transmembrane domain of AE1 are associated with dominant hereditary stomatocytosis, monovalent cation leaks, and reduced anion exchange activity. We set up a stopped-flow spectrofluorometry assay coupled with flow cytometry to investigate the anion transport and membrane expression characteristics of wild-type recombinant AE1 in HEK293 cells, using an inducible expression system. Likewise, study of three stomatocytosis-associated mutations (R730C, E758K, and G796R), allowed the validation of our method. Measurement of the rapid and specific chloride/bicarbonate exchange by surface expressed AE1 showed that E758K mutant was fully active compared with wild-type (WT) AE1, whereas R730C and G796R mutants were inactive, reinforcing previously reported data on other experimental models. Stopped-flow analysis of AE1 transport activity in red blood cell ghost preparations revealed a 50% reduction of G796R compared with WT AE1 corresponding to a loss of function of the G796R mutated protein, in accordance with the heterozygous status of the AE1 variant patients. In conclusion, stopped-flow led to measurement of rapid transport kinetics using the natural substrate for AE1 and, conjugated with flow cytometry, allowed a reliable correlation of chloride/bicarbonate exchange to surface expression of AE1, both in recombinant cells and ghosts and therefore a fine comparison of function between different stomatocytosis samples. This technical approach thus provides significant improvements in anion exchange analysis in red blood cells.