Merck
  • A putative human zinc-finger gene (ZFPL1) on 11q13, highly conserved in the mouse and expressed in exocrine pancreas. The European Consortium on MEN 1.
A putative human zinc-finger gene (ZFPL1) on 11q13, highly conserved in the mouse and expressed in exocrine pancreas. The European Consortium on MEN 1.
Genomics (1998-07-08)
J W Höppener, M J De Wit, A Y Simarro-Doorten, J F Roijers, H M van Herrewaarden, C J Lips, F Parente, D Quincey, P Gaudray, S Khodaei, G Weber, B Teh, F Farnebo, C Larsson, C X Zhang, A Calender, A A Pannett, S A Forbes, J H Bassett, R V Thakker, I Lemmens, W J Van de Ven, K Kas
RESUMO
In the process of identification of the multiple endocrine neoplasia type 1 gene, which was recently published, we isolated a novel gene in the 11q13 region. This gene (named ZFPL1, for zinc-finger protein-like 1) is expressed strongly in the exocrine pancreas as a 1.4-kb polyadenylated RNA encoding a putative protein of 310 amino acids. A mouse EST contig predicts an equally sized murine protein with 91% amino acid sequence identity to the human protein. No significant homology with known proteins could be found through database screening. However, zinc-finger-like domains and leucine-zipper-like motifs in the predicted ZFPL1 protein were identified, suggesting the presence of DNA-binding and dimerization domains possibly involved in transcription regulation. This notion is supported by the presence of a putative bipartite nuclear localization signal. This paper presents the full-length cDNA sequence for this gene, its genomic structure and chromosomal orientation, and expression studies by Northern blot hybridization and RNA in situ hybridization.