Merck
  • Highly efficient endosomal labeling of progenitor and stem cells with large magnetic particles allows magnetic resonance imaging of single cells.
Highly efficient endosomal labeling of progenitor and stem cells with large magnetic particles allows magnetic resonance imaging of single cells.
Blood (2003-04-05)
Kathleen A Hinds, Jonathan M Hill, Erik M Shapiro, Mikko O Laukkanen, Alfonso C Silva, Christian A Combs, Timothy R Varney, Robert S Balaban, Alan P Koretsky, Cynthia E Dunbar
RESUMO
Tracking transplanted stem cells using magnetic resonance imaging (MRI) could offer biologic insight into homing and engraftment. Ultrasmall dextran-coated iron oxide particles have previously been developed for uptake into cells to allow MRI tracking. We describe a new application of much larger, micron-scale, iron oxide magnetic particles with enhanced MR susceptibility, which enables detection of single cells at resolutions that can be achieved in vivo. In addition, these larger particles possess a fluorophore for histologic confirmation of cell distribution. We demonstrate highly efficient, nontoxic, endosomal uptake of these particles into hematopoietic CD34+ cells and mesenchymal stem cells documented by confocal and electron microscopy. Labeled cells retain biologic activity with preservation of colony-forming ability and differentiation capacity. MRI studies could detect labeled CD34+ cells and mesenchymal stem cells (MSCs) at single cell resolution. This appears to be a promising tool for serial noninvasive monitoring of in vivo cell homing and localization using MRI.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sodium cacodylate trihydrate, purum p.a., ≥98.0% (T)