Merck
  • Prominent expression and activity-dependent nuclear translocation of Ca2+/calmodulin-dependent protein kinase Idelta in hippocampal neurons.
Prominent expression and activity-dependent nuclear translocation of Ca2+/calmodulin-dependent protein kinase Idelta in hippocampal neurons.
The European journal of neuroscience (2005-12-06)
Hiroyuki Sakagami, Akifumi Kamata, Hideki Nishimura, Jiro Kasahara, Yuji Owada, Yusuke Takeuchi, Makoto Watanabe, Kohji Fukunaga, Hisatake Kondo
RESUMO
Multifunctional Ca2+/calmodulin-dependent protein kinases (CaMKs) including CaMKI, II and IV, are thought to regulate a variety of neuronal functions. Unlike CaMKII, which is regulated by autophosphorylation, CaMKI as well as CaMKIV are activated by CaMKK. In this study, we examined the cellular and subcellular localization of CaMKIdelta, a recently identified fourth isoform of CaMKI, in the mature brain. In situ hybridization analysis demonstrated wide expression of CaMKIdelta mRNA in the adult mouse brain with prominent expression in the hippocampal pyramidal cells. FLAG-tagged CaMKIdelta was localized at the cytoplasm and neurites without nuclear immunoreactivity in approximately 80% of the transfected primary hippocampal neurons. The stimulation with either KCl depolarization or glutamate triggered the nuclear localization of FLAG-tagged CaMKIdelta by two-fold with a peak at 1 min. In contrast, the catalytically inactive mutants of CaMKIdelta remained cytoplasmic without nuclear translocation during KCl depolarization, indicating the requirement of its activation for the nuclear translocation. Furthermore, we showed that immunoprecipitated CaMKIdelta could phosphorylate cAMP response element binding protein (CREB)alphain vitro and that the over-expression of CaMKIdelta enhanced GAL4-CREB-luciferase activity in PC12 cells stimulated by KCl depolarization. Our present study provides the first evidence for the possible involvement of CaMKIdelta in nuclear functions through its nuclear translocation in response to stimuli that trigger intracellular Ca2+ influx.