Merck
  • Isoprostane, an "intermediate phenotype" for oxidative stress heritability, risk trait associations, and the influence of chromogranin B polymorphism.
Isoprostane, an "intermediate phenotype" for oxidative stress heritability, risk trait associations, and the influence of chromogranin B polymorphism.
Journal of the American College of Cardiology (2010-10-05)
Fangwen Rao, Kuixing Zhang, Srikrishna Khandrika, Manjula Mahata, Maple M Fung, Michael G Ziegler, Brinda K Rana, Daniel T O'Connor
RESUMO
The purpose of this study is to understand whether isoprostane, a biomarker of oxidative stress, is subject to heritable control; whether it shares heritability with other cardiometabolic risk traits; and finally whether genetic variation at a specific candidate locus contributes to isoprostane variability. Isoprostane marks oxidative stress, and elevated isoprostane excretion might be involved in cardiovascular target organ damage. Here we used the classical twin pair method to probe the role of heredity in generating the isoprostane trait. Trait heritability (h(2)) and shared genetic determination among traits (pleiotropy, genetic covariance, ρ(G)) were estimated by variance components in twin pairs. Because the isoprostane and Chromogranin B (CHGB) traits shared ρ(G), we examined the CHGB locus for effects on the traits. Urinary isoprostane excretion was substantially heritable (h(2) = 65.8 ± 4.3%), and the isoprostane trait aggregated with multiple traits (CHGB, catecholamines, autonomic/baroreceptor, and renal function), including several features of the metabolic syndrome (body mass index, insulin resistance, dyslipidemia). Isoprostane excretion also aggregated with systemic hypertension. Twin studies demonstrated genetic covariance (pleiotropy) for the isoprostane and CHGB traits (ρ(G) = 0.27), and therefore we investigated the CHGB locus for trait effects. A common variant in the 3'-UTR of CHGB (C+84A) associated with plasma CHGB as well as isoprostane excretion. The C+84A disrupted an A/U-rich messenger ribonucleic acid stability element, and in transfected luciferase/3'-UTR plasmids, the C+84 and +84A alleles differed markedly in reporter expression in chromaffin and neuroblastoma cells, whereas site-directed mutagenesis confirmed the importance of this variant within the context of the A/U-rich motif. Isoprostane excretion is substantially heritable and shares joint genetic determination with CHGB as well as multiple features of the metabolic syndrome. A common polymorphism in the 3'-UTR (C+84A) of CHGB, which disrupts an A/U-rich messenger ribonucleic acid stability element, associates with not only CHGB secretion but also excretion of isoprostane. We propose a chain of events whereby CHGB genetic variation results in oxidative stress, with isoprostane formation. The results suggest novel links among the catecholaminergic system, oxidative pathways, and systemic hypertension.