Merck
Deletions in PITX1 cause a spectrum of lower-limb malformations including mirror-image polydactyly.
European journal of human genetics : EJHG (2012-01-20)
Eva Klopocki, Christian Kähler, Nicola Foulds, Hitesh Shah, Benjamin Joseph, Hermann Vogel, Sabine Lüttgen, Rainer Bald, Regina Besoke, Karsten Held, Stefan Mundlos, Ingo Kurth
RESUMO
PITX1 is a bicoid-related homeodomain transcription factor implicated in vertebrate hindlimb development. Recently, mutations in PITX1 have been associated with autosomal-dominant clubfoot. In addition, one affected individual showed a polydactyly and right-sided tibial hemimelia. We now report on PITX1 deletions in two fetuses with a high-degree polydactyly, that is, mirror-image polydactyly. Analysis of DNA from additional individuals with isolated lower-limb malformations and higher-degree polydactyly identified a third individual with long-bone deficiency and preaxial polydactyly harboring a heterozygous 35 bp deletion in PITX1. The findings demonstrate that mutations in PITX1 can cause a broad spectrum of isolated lower-limb malformations including clubfoot, deficiency of long bones, and mirror-image polydactyly.