Merck
  • Granulocyte macrophage-colony stimulating factor reduces the affinity of SHP-2 for the ITIM of CLECSF6 in neutrophils: a new mechanism of action for SHP-2.
Granulocyte macrophage-colony stimulating factor reduces the affinity of SHP-2 for the ITIM of CLECSF6 in neutrophils: a new mechanism of action for SHP-2.
Molecular immunology (2005-12-20)
Manon Richard, Nathalie Thibault, Patricia Veilleux, Geneviève Gareau-Pagé, André D Beaulieu
RESUMO
Proteins that bear immunoreceptor tyrosine based inhibitory motifs (ITIM) are believed to participate in the repression of cell activation via phosphatases such as SHP-1, SHP-2 and/or SHIP-1. CLECSF6, also called DCIR, is a transmembrane protein expressed on leukocytes and predominantly on neutrophils that bears one ITIM pattern. This feature confers to CLECSF6 a role in the repression of cell activation. In order to better understand its role in neutrophil signalling, we analysed the binding of phosphatases to the ITIM of CLECSF6. We showed that a peptide bearing the ITIM of CLECSF6 in its phosphorylated form associates with both SHP-1 and SHP-2. Phosphorylated SHP-1 binds the ITIM whereas phosphorylated SHP-2 does not. In addition, granulocyte macrophage-colony stimulating factor (GM-CSF) reduces the binding of SHP-2 to the ITIM of CLECSF6 while enhancing the phosphorylation level of SHP-2. GM-CSF is known to recruit SHP-2 to its receptor. These data suggest that the phosphorylation of SHP-2 by GM-CSF promotes the binding of SHP-2 to the GM-CSF receptor to the disadvantage of CLECSF6. Therefore, upon a treatment with GM-CSF, SHP-2 could move from a CLECSF6 associated signalosome with a repressor function to a GM-CSF receptor associated signalosome with an activator function.