Merck
Structure and chromosomal localization of the human glycogenin-2 gene GYG2.
Gene (2000-03-18)
L Zhai, J Mu, H Zong, A A DePaoli-Roach, P J Roach
RESUMO
Glycogenin-2 is one of two self-glucosylating proteins involved in the initiation phase of the synthesis of the storage polysaccharide glycogen. Cloning of the human glycogenin-2 gene, GYG2, has revealed the presence of 11 exons and a gene of more than 46 kb in size. The structure of the gene explains much of the observed diversity in glycogenin-2 cDNA sequences as being due to alternate exon usage. In some cases, there is variation in the splice junctions used. Over regions of protein sequence similarity, the GYG2 gene structure is similar to that of the other glycogenin gene, GYG. A genomic GYG2 clone was used to localize the gene to Xp22.3 by fluorescence in-situ hybridization. Localization close to the telomere of the short arm of the X chromosome is consistent with mapping information obtained from glycogenin-2 STS sequences. Glycogenin-2 maps between the microsatellite anchor markers AFM319te9 (DXS7100) and AFM205tf2 (DXS1060), and its 3' end is 34.5 kb from the 3' end of the arylsulphatase gene ARSD. GYG2 is outside the pseudoautosomal region PAR1 but still in a region of X-Y shared genes. As is true for several other genes in this location, an inactive remnant of GYG2, consisting of exons 1-3, may be present on the Y chromosome.