Merck
  • The BRCA1-associated protein BACH1 is a DNA helicase targeted by clinically relevant inactivating mutations.
The BRCA1-associated protein BACH1 is a DNA helicase targeted by clinically relevant inactivating mutations.
Proceedings of the National Academy of Sciences of the United States of America (2004-02-26)
Sharon Cantor, Ronny Drapkin, Fan Zhang, Yafang Lin, Juliana Han, Sushmita Pamidi, David M Livingston
RESUMO
BACH1 is a nuclear protein that directly interacts with the highly conserved, C-terminal BRCT repeats of the tumor suppressor, BRCA1. Mutations within the BRCT repeats disrupt the interaction between BRCA1 and BACH1, lead to defects in DNA repair, and result in breast and ovarian cancer. BACH1 is necessary for efficient double-strand break repair in a manner that depends on its association with BRCA1. Moreover, some women with early-onset breast cancer and no abnormalities in either BRCA1 or BRCA2 carry germline BACH1 coding sequence changes, suggesting that abnormal BACH1 function contributes to tumor induction. Here, we show that BACH1 is both a DNA-dependent ATPase and a 5'-to-3' DNA helicase. In two patients with early-onset breast cancer who carry distinct germline BACH1 coding sequence changes, the resulting proteins are defective in helicase activity, indicating that these sequence changes disrupt protein function. These results reinforce the notion that mutant BACH1 participates in breast cancer development.