Merck
Mitochondrial thioredoxin reductase is essential for early postischemic myocardial protection.
Circulation (2011-12-07)
Jan Horstkotte, Tamara Perisic, Manuela Schneider, Philipp Lange, Melanie Schroeder, Claudia Kiermayer, Rabea Hinkel, Tilman Ziegler, Pankaj K Mandal, Robert David, Sabine Schulz, Sabine Schmitt, Julian Widder, Fred Sinowatz, Bernhard F Becker, Johann Bauersachs, Michael Naebauer, Wolfgang M Franz, Irmela Jeremias, Markus Brielmeier, Hans Zischka, Marcus Conrad, Christian Kupatt
RESUMO
Excessive formation of reactive oxygen species contributes to tissue injury and functional deterioration after myocardial ischemia/reperfusion. Especially, mitochondrial reactive oxygen species are capable of opening the mitochondrial permeability transition pore, a harmful event in cardiac ischemia/reperfusion. Thioredoxins are key players in the cardiac defense against oxidative stress. Mutations in the mitochondrial thioredoxin reductase (thioredoxin reductase-2, Txnrd2) gene have been recently identified to cause dilated cardiomyopathy in patients. Here, we investigated whether mitochondrial thioredoxin reductase is protective against myocardial ischemia/reperfusion injury. In mice, α-MHC-restricted Cre-mediated Txnrd2 deficiency, induced by tamoxifen (Txnrd2-/-ic), aggravated systolic dysfunction and cardiomyocyte cell death after ischemia (90 minutes) and reperfusion (24 hours). Txnrd2-/-ic was accompanied by a loss of mitochondrial integrity and function, which was resolved on pretreatment with the reactive oxygen species scavenger N-acetylcysteine and the mitochondrial permeability transition pore blocker cyclosporin A. Likewise, Txnrd2 deletion in embryonic endothelial precursor cells and embryonic stem cell-derived cardiomyocytes, as well as introduction of Txnrd2-shRNA into adult HL-1 cardiomyocytes, increased cell death on hypoxia and reoxygenation, unless N-acetylcysteine was coadministered. We report that Txnrd2 exerts a crucial function during postischemic reperfusion via thiol regeneration. The efficacy of cyclosporin A in cardiac Txnrd2 deficiency may indicate a role for Txnrd2 in reducing mitochondrial reactive oxygen species, thereby preventing opening of the mitochondrial permeability transition pore.