Merck
  • A novel vitronectin receptor integrin (alpha v beta x) is responsible for distinct adhesive properties of carcinoma cells.
A novel vitronectin receptor integrin (alpha v beta x) is responsible for distinct adhesive properties of carcinoma cells.
Cell (1989-04-07)
D A Cheresh, J W Smith, H M Cooper, V Quaranta
RESUMO
Carcinoma cells express a novel integrin involved in cell adhesion to vitronectin, but not to fibrinogen or von Willebrand factor, whereas melanoma and endothelial cells express a vitronectin receptor (alpha v beta 3) that promotes cell attachment to all of these matrix components. The integrin responsible for this adhesive phenotype of carcinoma cells is composed of an alpha subunit that is indistinguishable from the alpha v of the vitronectin receptor and a beta subunit (beta x) that is distinct from any known integrin beta subunit. Accordingly, Northern blot analysis identifies an mRNA for alpha v, but not for beta 3 in carcinoma cells. This receptor appears to mediate cell adhesion to vitronectin as well as fibronectin since an antibody directed to its alpha subunit blocked carcinoma cell adhesion to both of these matrix proteins. These results suggest that homologous integrins with identical alpha subunits and structurally distinct beta subunits can account for the functional recognition of different matrixes by two cell types.