Merck
  • Synthesis, structural characterization, and thermoresponsivity of hybrid supramolecular dendrimers bearing a polyoxometalate core.
Synthesis, structural characterization, and thermoresponsivity of hybrid supramolecular dendrimers bearing a polyoxometalate core.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-07-03)
Hailong Chen, Yang Yang, Yizhan Wang, Lixin Wu
RESUMO
A series of cationic dendrons bearing triethylene glycol monomethyl ether terminal groups of different generations have been synthesized and used to encapsulate an inorganic polyanionic cluster [K(12.5)Na(1.5)(NaP5W30O110)] through electrostatic interactions. The resulting dendritic cation-encapsulated polyoxometalate (POM) complexes, cluster-dendrimers, are soluble in water and exhibit lower critical solution temperatures (LCST). The thermoresponsivities of these complexes in aqueous solutions were studied by turbidimetry and variable-temperature (1)H NMR spectroscopy. The observed cloud points show a remarkable dependence on the generation of the dendrons. Complexes composed of first-generation dendrons exhibit no obvious thermoresponsive properties, but for complexes bearing second-generation dendrons, the LCST decreases as the number of dendritic cations around the POM cluster increases. Complexes composed of third-generation cations underwent reversible aggregation and disaggregation upon heating and cooling, respectively. This thermally induced self-aggregation was characterized by DLS and TEM. In addition, the effects of salt and solvent on the LCST were investigated. This research demonstrates a new type of thermoresponsive dendritic organic-inorganic hybrid complex and provides a general route to the endowment of POMs with temperature-sensitive properties through electrostatic interactions.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triethylene glycol monomethyl ether, purum, ≥97.0% (GC)