Merck
A direct role of Mad1 in the spindle assembly checkpoint beyond Mad2 kinetochore recruitment.
EMBO reports (2014-01-31)
Thomas Kruse, Marie Sofie Yoo Larsen, Garry G Sedgwick, Jón Otti Sigurdsson, Werner Streicher, Jesper V Olsen, Jakob Nilsson
RESUMO
The spindle assembly checkpoint (SAC) ensures accurate chromosome segregation by delaying entry into anaphase until all sister chromatids have become bi-oriented. A key component of the SAC is the Mad2 protein, which can adopt either an inactive open (O-Mad2) or active closed (C-Mad2) conformation. The conversion of O-Mad2 into C-Mad2 at unattached kinetochores is thought to be a key step in activating the SAC. The "template model" proposes that this is achieved by the recruitment of soluble O-Mad2 to C-Mad2 bound at kinetochores through its interaction with Mad1. Whether Mad1 has additional roles in the SAC beyond recruitment of C-Mad2 to kinetochores has not yet been addressed. Here, we show that Mad1 is required for mitotic arrest even when C-Mad2 is artificially recruited to kinetochores, indicating that it has indeed an additional function in promoting the checkpoint. The C-terminal globular domain of Mad1 and conserved residues in this region are required for this unexpected function of Mad1.