Merck
LCK is an important mediator of B-cell receptor signaling in chronic lymphocytic leukemia cells.
Molecular cancer research : MCR (2013-03-19)
Fatima Talab, John C Allen, Victoria Thompson, Ke Lin, Joseph R Slupsky
RESUMO
B-cell receptor (BCR) signals promote survival of chronic lymphocytic leukemia (CLL) cells, and it is believed that overexpressed and constitutively active Lyn mediates this signaling. Here, we show that CLL cells express lymphocyte-specific protein tyrosine kinase (LCK) and that inhibition of this Src family tyrosine kinase with the specific inhibitor [4-amino-5-(4-phenoxyphenyl)-7H-pyrrolo[3,2-d]pyrimidin-7-yl-cyclopentane (Lck-i)], or reduction of its expression with siRNA, blocks the induction of CD79a, Syk, inhibitor of IκB kinase (IKK), Akt, and extracellular signal-regulated kinase (ERK) phosphorylation by BCR cross-linking in these cells. Furthermore, we show that CLL cells with high levels of LCK expression have higher levels of BCR-mediated IKK, Akt, and ERK phosphorylation as well as cell survival than CLL cells with low levels of LCK expression. We also show that treatment of CLL cells with Lck-i inhibits BCR cross-linking-induced cell survival. Taken together, these data show a major role for LCK in proximal and distal BCR-mediated signaling in CLL cells and suggest that LCK expression is important in the pathogenesis of this disease. On a clinical level, these studies advocate the use of specific LCK inhibitors in the treatment of progressive CLL.