Merck
  • [Identification of transthyretin posttranslational modifications 1n human blood using mass-spectrometric methods].
[Identification of transthyretin posttranslational modifications 1n human blood using mass-spectrometric methods].
Molekuliarnaia biologiia (2014-12-17)
I A Popov, N L Starodubtseva, M I Indeĭkina, Iu I Kostiukevich, A S Kononikhin, M I Nikolaeva, E N Kukaev, S A Kozin, A A Makarov, E N Nikolaev
RESUMO
Transthyretin, one of the major plasma proteins, has a number of posttranslational modifications and mutations, some of which are associated with the development of severe diseases, for instance, familial amyloid neuropathy and Alzheimer's disease. In order to investigate the role of modified forms in the development of these diseases a complex analytical platform, based on two mass-spectrometric approaches (bottom-up and op-down) has been developed. The high efficiency of this method was shown using 10 plasma samples obtained from patients with Alzheimer's disease and healthy individuals.