Merck
  • Immunohistochemistry in apparently normal bone marrow trephine specimens from patients with nodal follicular lymphoma.
Immunohistochemistry in apparently normal bone marrow trephine specimens from patients with nodal follicular lymphoma.
Journal of clinical pathology (1995-11-01)
R Chetty, G Echezarreta, M Comley, K Gatter
RESUMO
To establish the role of immunohistochemistry (using a limited panel of antibodies) in detecting minimal involvement by follicular lymphoma in routinely processed bone marrow trephine specimens, which show no obvious morphological (light microscopic) evidence of lymphoma; to determine whether bcl-2 immunostaining in bone marrow distinguishes between benign and malignant infiltrates in a patient with nodal follicular lymphoma. Twenty seven consecutively selected paraffin wax embedded, formalin fixed bone marrow trephine specimens were stained with the following antibodies: anti-bcl-2, anti-CD79a, anti-CD3, and kappa and lambda light chains, using the Streptavidin biotin complex technique. Five of the 27 cases, which showed no evidence of involvement by follicular lymphoma on routine stains, showed monotypic B cells on immunohistochemistry. Two of the cases were diffuse, while the remaining three showed mini-aggregates around bony trabeculae. In all five cases the lymphomatous infiltrates were strongly bcl-2 positive. Reactive B lymphoid nodules did not show the same degree of bcl-2 positivity, and negative cells could be discerned within the reactive nodules. There is merit in studying so-called negative bone marrows immunohistochemically in order to detect minimal involvement by follicular lymphoma. A limited panel of antibodies including anti-bcl-2, anti-CD79a and anti-CD3 is usually adequate to accomplish this. Strongly bcl-2 positive lymphoid aggregates in the bone marrow of patients with nodal follicular lymphoma are indicative of lymphoma.