Merck
High-performance air-stable organic field-effect transistors: isoindigo-based conjugated polymers.
Journal of the American Chemical Society (2011-04-07)
Ting Lei, Yue Cao, Yunlong Fan, Chen-Jiang Liu, Si-Chun Yuan, Jian Pei
RESUMO
Two conjugated polymers, IIDDT and IIDT, based on an isoindigo core were developed for organic field-effect transisitors. Investigation of their field-effect performance indicated that IIDDT exhibited air-stable mobility up to 0.79 cm(2) V(-1) s(-1), which is quite high among polymer FET materials. The facile preparation and high mobility of such polymers make isoindigo-based polymers very promising for application as solution-processable organic semiconductors for optoelectronic devices.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
6,6′-Dibromoisoindigo
Preço e disponibilidade não estão disponíveis no momento.