Merck
Pyrene-modified PNAs: Stacking interactions and selective excimer emission in PNA2DNA triplexes.
Beilstein journal of organic chemistry (2014-08-28)
Alex Manicardi, Lucia Guidi, Alice Ghidini, Roberto Corradini
RESUMO
Pyrene derivatives can be incorporated into nucleic acid analogs in order to obtain switchable probes or supramolecular architectures. In this paper, peptide nucleic acids (PNAs) containing 1 to 3 1-pyreneacetic acid units (PNA1-6) with a sequence with prevalence of pyrimidine bases, complementary to cystic fibrosis W1282X point mutation were synthesized. These compounds showed sequence-selective switch-on of pyrene excimer emission in the presence of target DNA, due to PNA2DNA triplex formation, with stability depending on the number and positioning of the pyrene units along the chain. An increase in triplex stability and a very high mismatch-selectivity, derived from combined stacking and base-pairing interactions, were found for PNA2, bearing two distant pyrene units.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1-Pyreneacetic acid, 97%