Merck
Stx5 is a novel interactor of VLDL-R to affect its intracellular trafficking and processing.
Experimental cell research (2013-05-25)
Timo Wagner, Marco Dieckmann, Sebastian Jaeger, Sascha Weggen, Claus U Pietrzik
RESUMO
We identified syntaxin 5 (Stx5), a protein involved in intracellular vesicle trafficking, as a novel interaction partner of the very low density lipoprotein (VLDL)-receptor (VLDL-R), a member of the LDL-receptor family. In addition, we investigated the effect of Stx5 on VLDL-R maturation, trafficking and processing. Here, we demonstrated mutual association of both proteins using several in vitro approaches. Furthermore, we detected a special maturation phenotype of VLDL-R resulting from Stx5 overexpression. We found that Stx5 prevented advanced Golgi-maturation of VLDL-R, but did not cause accumulation of the immature protein in ER, ER to Golgi compartments, or cis-Golgi ribbon, the main expression sites of Stx5. Rather more, abundantly present Stx5 was capable of translocating ER-/N-glycosylated VLDL-R to the plasma membrane, and thus was insensitive to BFA treatment and low temperature. Furthermore, abundant presence of Stx5 significantly interfered with VLDL-R reaching the trans-Golgi network. Based on our findings, we postulate that Stx5 can directly bind to the C-terminal domain of VLDL-R, thereby influencing the receptor's glycosylation, trafficking and processing characteristics. Resulting from that, we further suggest that Stx5 might play a role in modulating VLDL-R physiology by participating in an abrasively described or completely novel Golgi-bypass pathway.