Merck
One-pot synthesis of functionalized spirobenzofuranones via MCR involving 3-cyanochromones.
The Journal of organic chemistry (2011-10-14)
Tryfon Zarganes-Tzitzikas, Michael A Terzidis, Julia Stephanidou-Stephanatou, Constantinos A Tsoleridis, George E Kostakis
RESUMO
Another aspect concerning chromone chemistry leading to the one-pot synthesis of functionalized novel spirobenzofuranones has been described. The synthesis involves reaction of the zwitterionic intermediates formed by the 1:1 interaction between isocyanides and acetylenecarboxylates with 3-cyanochromones, whereupon through an unexpected and unprecedented reaction of the chromone moiety the isolated benzofuranones are formed. The regioselectivity of the reaction was investigated by DFT calculations. The geometries of the intermediates, transition structures, and intermediate products, leading to the final products, were optimized using the B3LYP functional with the 6-31G(d) basis set. The structures of the products were elucidated by 1D and 2D NMR experiments. Full assignment of all (1)H and (13)C NMR chemical shifts has been achieved. A plausible mechanistic rationale is proposed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-Cyanochromone, 97%