Merck
  • Endocannabinoid biosynthesis proceeding through glycerophospho-N-acyl ethanolamine and a role for alpha/beta-hydrolase 4 in this pathway.
Endocannabinoid biosynthesis proceeding through glycerophospho-N-acyl ethanolamine and a role for alpha/beta-hydrolase 4 in this pathway.
The Journal of biological chemistry (2006-07-05)
Gabriel M Simon, Benjamin F Cravatt
RESUMO
N-Acyl ethanolamines (NAEs) are a large class of signaling lipids implicated in diverse physiological processes, including nociception, cognition, anxiety, appetite, and inflammation. It has been proposed that NAEs are biosynthesized from their corresponding N-acyl phosphatidylethanolamines (NAPEs) in a single enzymatic step catalyzed by a phospholipase D (NAPE-PLD). The recent generation of NAPE-PLD(-/-) mice has revealed that these animals possess lower brain levels of saturated NAEs but essentially unchanged concentrations of polyunsaturated NAEs, including the endogenous cannabinoid anandamide. These findings suggest the existence of additional enzymatic routes for the production of NAEs in vivo. Here, we report evidence for an alternative pathway for NAE biosynthesis that proceeds through the serine hydrolase-catalyzed double-deacylation of NAPE to generate glycerophospho-NAE, followed by the phosphodiesterase-mediated cleavage of this intermediate to liberate NAE. Furthermore, we describe the functional proteomic isolation and identification of a heretofore uncharacterized enzyme alpha/beta-hydrolase 4 (Abh4) as a lysophospholipase/phospholipase B that selectively hydrolyzes NAPEs and lysoNAPEs. Abh4 accepts lysoNAPEs bearing both saturated and polyunsaturated N-acyl chains as substrates and displays a distribution that closely mirrors lysoNAPE-lipase activity in mouse tissues. These results support the existence of an NAPE-PLD-independent route for NAE biosynthesis and suggest that Abh4 plays a role in this metabolic pathway by acting as a (lyso)NAPE-selective lipase.
MATERIAIS
Número do produto
Marca
Descrição do produto
Avanti
AEA-d4, C20:4 anandamide-d4, methanol solution