Merck
  • Preparation, Purification, and Characterization of Binuclear Ruthenium(II) Complexes: Bridging Ligands Based on Diazafluorenes.
Preparation, Purification, and Characterization of Binuclear Ruthenium(II) Complexes: Bridging Ligands Based on Diazafluorenes.
Inorganic chemistry (2001-10-24)
Youxiang Wang, Willie J. Perez, Greg Y. Zheng, D. Paul Rillema, Connie L. Huber
RESUMO
A series of bimetallic complexes of ruthenium(II) bridged by heterocyclic ligands formed by the condensation of 4,5-diazafluoren-9-one with various diamines, hydrazine, 1,4-phenylenediamine, benzidine, and 4,4'-methylenedianiline, results in metal centers separated by various distances. The complexes give rise to metal-to-ligand charge-transfer absorptions in the 450 nm region of the visible spectrum and intraligand pi --> pi transitions in the 300 nm region of the ultraviolet spectrum. The ruthenium(II) centers are oxidized in two closely spaced one-electron processes at potentials more positive than that for Ru(bpy)(3)(2+). The bridging ligands are reduced by two closely spaced one-electron processes at more positive potentials than that for reduction of the coordinated bipyridine ligands ( approximately -1.30 V), although the diazafluorene=N-N=diazafluorene bridging ligand is reduced reversibly in two single-electron steps at E(1/2) values of -0.29 and -0.52 V. After purification by eluting the "pure" complexes over a silica gel column with various concentrations of 0.10 M NH(4)PF(6) in methanol, emission occurs only in a glassy matrix at 77 K and at low temperatures in solution. The emission lifetimes at 77 K in a 4:1 ethanol:methanol glass are 5 +/- 1 &mgr;s. A variable-temperature emission lifetime study reveals the presence of a low-lying state with DeltaE = approximately 1500 cm(-)(1), and extrapolation to room temperature indicates the emission lifetimes are in the subnanosecond range.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4,5-Diazafluoren-9-one, 97%