Merck
  • Biochemical characterization of two functional human liver acyl-CoA oxidase isoforms 1a and 1b encoded by a single gene.
Biochemical characterization of two functional human liver acyl-CoA oxidase isoforms 1a and 1b encoded by a single gene.
Biochemical and biophysical research communications (2007-07-03)
David Oaxaca-Castillo, Pierre Andreoletti, Aurore Vluggens, Sangtao Yu, Paul P van Veldhoven, Janardan K Reddy, Mustapha Cherkaoui-Malki
RESUMO
Human acyl-CoA oxidase 1 (ACOX1) is a rate-limiting enzyme in peroxisomal fatty acids beta-oxidation and its deficiency is associated with a lethal, autosomal recessive disease, called pseudoneonatal-adrenoleukodystrophy. Two mRNA variants, transcribed from a single gene encode ACOX1a or ACOX1b isoforms, respectively. Recently, a mutation in a splice site has been reported [H. Rosewich, H.R. Waterham, R.J. Wanders, S. Ferdinandusse, M. Henneke, D. Hunneman, J. Gartner, Pitfall in metabolic screening in a patient with fatal peroxisomal beta-oxidation defect, Neuropediatrics 37 (2006) 95-98.], which results in the defective peroxisomal fatty acids beta-oxidation. Here, we show that these mRNA splice variants are expressed differentially in human liver. We investigated the biochemical role of the two human ACOX1 isoforms by heterologous expression of the catalytically active ACOX1a and ACOX1b enzymes in Escherichia coli. ACOX1a seems to be more labile and exhibits only 50% specific activity toward palmitoyl-CoA as compared to ACOX1b.