Merck
Solid-phase synthesis of acridine-based threading intercalator peptides.
Bioorganic & medicinal chemistry letters (2000-09-15)
C B Carlson, P A Beal
RESUMO
The preparation of a novel acridine-based amino acid is reported. This N-Alloc-protected monomer can be coupled and deprotected under solid-phase peptide synthesis procedures to create acridine peptide conjugates as potential threading intercalators. A peptide containing this novel amino acid undergoes spectral changes in the presence of duplex DNA and RNA consistent with intercalative binding.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-Aminobenzylamine, 99%