Merck
  • {[Eth-yl(pyridin-4-ylmeth-yl)carbamo-thio-yl]sulfanido-κ(2) S,S'}(1,4,7,10,13,16-hexa-oxa-cyclo-octa-decane-κ(6) O)potassium.
{[Eth-yl(pyridin-4-ylmeth-yl)carbamo-thio-yl]sulfanido-κ(2) S,S'}(1,4,7,10,13,16-hexa-oxa-cyclo-octa-decane-κ(6) O)potassium.
Acta crystallographica. Section E, Structure reports online (2014-01-16)
Hadi D Arman, Pavel Poplaukhin, Edward R T Tiekink
RESUMO
The asymmetric unit of title salt co-crystal, [K(C9H11N2S2)(C12H24O6)], comprises a K(+) cation, an (-)S2CN(Et)py anion and a 18-crown-6 mol-ecule. Substantial delocalization of π-electron density is evident in the di-thio-carbamate anion, as indicated by the equivalent C-S bond lengths. The K(+) cation sits within an O6S2 donor set lying 0.7506 (6) Å out of the least-squares plane through the six O atoms (r.m.s. deviation = 0.1766 Å) of the 18-crown-6 mol-ecule with the two S atoms being on one side of this plane. Supra-molecular layers in the bc plane, sustained by C-H⋯O and C-H⋯π inter-actions, feature in the crystal packing.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-(Ethylaminomethyl)pyridine, 97%