Merck
  • Presence and type of low density lipoprotein receptor (LDLR) mutation influences the lipid profile and response to lipid-lowering therapy in Brazilian patients with heterozygous familial hypercholesterolemia.
Presence and type of low density lipoprotein receptor (LDLR) mutation influences the lipid profile and response to lipid-lowering therapy in Brazilian patients with heterozygous familial hypercholesterolemia.
Atherosclerosis (2014-02-18)
Paulo Caleb Junior Lima Santos, Aline Cruz Morgan, Cintia Elin Jannes, Luciana Turolla, Jose Eduardo Krieger, Raul D Santos, Alexandre Costa Pereira
RESUMO
Familial hypercholesterolemia (FH) is an autosomal dominant disease caused mainly by LDLR mutations. This study assessed the influence of the presence and type of LDLR mutation on lipid profile and the response to lipid-lowering therapy in Brazilian patients with heterozygous FH. For 14 ± 3 months, 156 patients with heterozygous FH receiving atorvastatin were followed. Coding sequences of the LDLR gene were bidirectionally sequenced, and the type of LDLR mutations were classified according to their probable functional class. The frequencies of the types of LDLR mutations were: null-mutation (n = 40, 25.6%), defective-mutation (n = 59, 37.8%), and without an identified mutation (n = 57, 36.6%). Baseline total cholesterol (TC) and low-density lipoprotein cholesterol (LDL-C) were higher in patients carrying a null mutation (9.9 ± 1.9 mmol/L, 7.9 ± 1.7 mmol/L), compared to those with a defective (8.9 ± 2.2 mmol/L, 7.0 ± 2.0 mmol/L), or no mutation (7.9 ± 1.9 mmol/L, 5.8 ± 1.9 mmol/L) (p < 0.001). After treatment, the proportion of patients attaining an LDL-C<3.4 mmol/L was significantly different among groups: null (22.5%), defective (27.1%), and without mutations (47.4%) (p = 0.02). The presence of LDLR mutations was independently associated with higher odds of not achieving the LDL-C cut-off (OR 9.07, 95% CI 1.41-58.16, p = 0.02). Our findings indicate that the presence and type of LDLR mutations influence lipid profile and response to lipid-lowering therapy in Brazilian patients with heterozygous FH. Thus, more intensive care with pharmacological therapeutics should be performed in patients who have a molecular analysis indicating the presence of a LDLR mutation.