Merck
  • Cloning and characterization of a human Mac-2-binding protein, a new member of the superfamily defined by the macrophage scavenger receptor cysteine-rich domain.
Cloning and characterization of a human Mac-2-binding protein, a new member of the superfamily defined by the macrophage scavenger receptor cysteine-rich domain.
The Journal of biological chemistry (1993-07-05)
K Koths, E Taylor, R Halenbeck, C Casipit, A Wang
RESUMO
We have purified and sequenced a secreted glycoprotein from both the human breast carcinoma cell line, SK-BR-3, and human breast milk. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2. This Mac-2 binding protein (Mac-2-BP) has an apparent native molecular mass of several million daltons and contains subunits of 85-97 kDa that are very susceptible to proteolysis at a dibasic cleavage site. Western analysis suggests that Mac-2-BP is found in serum, semen, saliva, urine, and tears, in addition to breast milk. The gene encoding Mac-2-BP was cloned from a cDNA bank of a human monocytic cell line, using degenerate PCR primers based on the protein sequence. Recombinant Mac-2-BP was expressed in Cos cells and secreted as a high molecular weight complex. The cDNA clone encodes a mature protein of 567 amino acids, preceded by an 18-amino acid leader. The mature protein contains 16 cysteines and has seven potential N-linked glycosylation sites. The first 106 amino acids represent a domain that is highly similar to an ancient protein superfamily defined by the macrophage scavenger receptor cysteine-rich domain.