Merck
  • Biodesulfurization of dibenzothiophene and its derivatives through the selective cleavage of carbon-sulfur bonds by a moderately thermophilic bacterium Bacillus subtilis WU-S2B.
Biodesulfurization of dibenzothiophene and its derivatives through the selective cleavage of carbon-sulfur bonds by a moderately thermophilic bacterium Bacillus subtilis WU-S2B.
Journal of bioscience and bioengineering (2005-10-20)
K Kirimura, T Furuya, Y Nishii, Y Ishii, K Kino, S Usami
RESUMO
Heterocyclic organosulfur compounds such as dibenzothiophene (DBT) in petroleum cannot be completely removed by hydrodesulfurization using chemical catalysts. A moderately thermophilic bacterium Bacillus subtilis WU-S2B, which could desulfurize DBT at 50 degrees C through the selective cleavage of carbon-sulfur (CS) bonds, was newly isolated. At 50 degrees C, growing cells of WU-S2B could degrade 0.54 mM DBT within 120 h to produce 2-hydroxybiphenyl, and the resting cells could also degrade 0.81 mM DBT within 12 h. The DBT-desulfurizing ability of WU-S2B is high over a wide temperature range from 30 to 50 degrees C, and highest at 50 degrees C for both the growing and resting cells, and this is an extremely advantageous property for the practical biodesulfurization. In addition, WU-S2B could also desulfurize DBT derivatives such as 2,8-dimethylDBT, 4,6-dimethylDBT and 3,4-benzoDBT. Therefore, B. subtilis WU-S2B is considered to have more beneficial properties than other desulfurizing bacteria such as Rhodococcus strains previously reported, particularly from the viewpoint of its capacity for thermophilic desulfurization through the CS bond cleavage.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dibenzothiophene, ≥99%