Merck
  • Design and synthesis of iridium(III) azacrown complex: application as a highly sensitive metal cation phosphorescence sensor.
Design and synthesis of iridium(III) azacrown complex: application as a highly sensitive metal cation phosphorescence sensor.
Organic & biomolecular chemistry (2005-12-17)
Mei-Lin Ho, Fu-Ming Hwang, Pei-Nung Chen, Ya-Hui Hu, Yi-Ming Cheng, Kung-Shih Chen, Gene-Hsiang Lee, Yun Chi, Pi-Tai Chou
RESUMO
A new metal cation probe bearing a central Ir(III) element and 1-aza-15-crown-5-ether substituted pyridyl pyrazolate as the chelate was synthesized. The octahedral molecular structure of was confirmed using single crystal X-ray diffraction analyses. Subsequent photophysical study showed yellow-green emission at approximately 560 nm in both fluid solution and solid state at room temperature. Remarkable differentiation in spectral properties upon metal cation (e.g. Ca2+) complexation makes complex a highly sensitive phosphorescence probe.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triethylene glycol di(p-toluenesulfonate), 98%