- Clobetasol propionate spray 0.05% for the treatment of moderate to severe plaque psoriasis.
Clobetasol propionate spray 0.05% for the treatment of moderate to severe plaque psoriasis.
Cutis (2012-04-06)
Alan Menter
PMID22474733
RESUMO
Clobetasol propionate is a super-high potent class 1 topical corticosteroid available in several formulations, including a spray formulation that is approved for use up to 4 weeks in patients aged 18 years and older with moderate to severe plaque psoriasis. The efficacy and safety of clobetasol propionate spray 0.05% has been extensively evaluated in clinical trials in more than 2200 patients with moderate to severe plaque psoriasis. This article reviews the efficacy, safety, and tolerability of clobetasol propionate spray 0.05%. Clobetasol propionate spray 0.05% is a topical product with a documented efficacy and safety profile with good acceptability in patients with moderate to severe plaque psoriasis.