Merck
  • UCN-01 (7-hydroxystaurosporine) and other indolocarbazole compounds: a new generation of anti-cancer agents for the new century?
UCN-01 (7-hydroxystaurosporine) and other indolocarbazole compounds: a new generation of anti-cancer agents for the new century?
Anti-cancer drug design (2000-07-11)
S Akinaga, K Sugiyama, T Akiyama
RESUMO
UCN-01 (7-hydroxystaurosporine) is a protein kinase inhibitor which is under development as an anti-cancer agent in the USA and Japan. Although UCN-01 was originally isolated from the culture broth of Streptomyces sp. as a protein kinase C-selective inhibitor, its ultimate target as an anti-cancer agent remains elusive. As a single agent, UCN-01 exhibits two key biochemical effects, namely accumulation of cells in the G1 phase of the cell cycle and induction of apoptosis. Both these effects may be important for its anti-cancer activity. As a modulator, UCN-01 enhances the cytotoxicity of other anti-cancer drugs such as DNA-damaging agents and anti-metabolite drugs through putative abrogation of G2 and/or S phase accumulation induced by these anti-cancer agents. Currently, in addition to UCN-01, four other indolocarbazole anti-cancer drugs-two protein kinase inhibitors, CGP 41251, CEP-751, and two DNA-damaging agents, NB-506 and a Rebeccamycin analog-are undergoing clinical investigations in the USA, Europe or Japan. In this review, we would like to address the differences and similarities of these indolocarbazole compounds as anti-cancer agents with regard to their mechanism(s) of action, the effects on cell cycle progression, induction of apoptosis and modulation of drug sensitivity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
UCN-01, ≥97% (HPLC), powder