Merck
  • Novel Bernard-Soulier syndrome variants caused by compound heterozygous mutations (case I) or a cytoplasmic tail truncation (case II) of GPIbα.
Novel Bernard-Soulier syndrome variants caused by compound heterozygous mutations (case I) or a cytoplasmic tail truncation (case II) of GPIbα.
Thrombosis research (2013-02-19)
Naomasa Yamamoto, Noriko Akamatsu, Hitoshi Sakuraba, Kazuhiko Matsuno, Ryota Hosoya, Haruo Nogami, Kohji Kasahara, Susumu Mitsuyama, Morio Arai
RESUMO
A defective platelet glycoprotein (GP) Ib/IX/V complex [von Willebrand factor (VWF) receptor] results in Bernard-Soulier syndrome (BSS), which is characterized by macrothrombocytopenia and impaired ristocetin- and thrombin-induced platelet aggregation. We found 2 independent BSS-variant families: Case I [compound heterozygous mutations, p.Glu331X and a frame shift by a deletion at c.1444delA of GPIbα (GP1BA) terminating at a premature stop codon (p.Thr452ProfsX58)], and case II [homozygous nonsense mutation at c.1723C>T, p.Gln545X]. Case I platelets expressed no GPIbα, resulting in absence of ristocetin-induced platelet aggregation (RIPA) and 50% reduction in thrombin-induced aggregation with no shape change. The mother's platelets had 50% the expression level of A-type GPIbα (4-repeated VNTR: variable number of tandem repeats, p.[Thr145Met; Ser399_Pro411[4]]); the father's platelets had the same expression level of C-type GPIbα (2-repeated VNTR, p.Ser399_Pro411dup) as the mother's platelets. The mother's RIPA was significantly higher than the father's. Thrombin-induced aggregation was normal in both parents. Case II platelets expressed a GPIbα with an abnormal cytoplasmic tail, p.Gln545X-truncated GPIbα, which complexed with GPIX and GPV on the cell surface; its expression level of the complex was normal. Case II platelets had reversible RIPA, with no ATP release, and weak thrombin-induced aggregation without shape change. These results suggest that a signaling process through the GPIbα cytoplasmic tail required for full platelet activation is defective in BSS variant case II and a length polymorphism of GPIbα is associated with a modified level of RIPA heterozygous BSS case I.