Merck
  • Synthesis of heterotelechelic poly(ethylene glycol) derivatives having alpha-benzaldehyde and omega-pyridyl disulfide groups by ring opening polymerization of ethylene oxide using 4-(diethoxymethyl)benzyl alkoxide as a novel initiator.
Synthesis of heterotelechelic poly(ethylene glycol) derivatives having alpha-benzaldehyde and omega-pyridyl disulfide groups by ring opening polymerization of ethylene oxide using 4-(diethoxymethyl)benzyl alkoxide as a novel initiator.
Bioconjugate chemistry (2004-03-18)
Yoshitsugu Akiyama, Yukio Nagasaki, Kazunori Kataoka
RESUMO
New heterotelechelic PEG-containing benzaldehyde and 2-pyridyldithio endgroup (CHO-Bz-PEG-SSpyl) was synthesized with high efficiency and high selectivity. An alpha-benzylacetal-omega-methansulfonyl PEG was prepared as the first step to CHO-Bz-PEG-SSpyl through the ring-opening polymerization of ethylene oxide (EO) initiated by potassium 4-(diethoxymethyl)benzyl alkoxide (PDA), followed by the successive conversion of the end-alkoxide group to a methanesulfonyl group and then to dithiocarbonate derivative. Further, deprotection of the dithiocarbonate derivative and subsequent conversion to the 2-pyridyldithio group at the omega-end was successfully performed through a one-step reaction to form alpha-benzylacetal-omega-2-pyridyldithio PEG (aceBz-PEG-SSpyl). The aceBz-PEG-SSpyl was then treated with an aqueous HCl solution (pH 5.0) to generate the benzaldehyde group at the alpha-end. Molecular functionalities of the benzaldehyde and the 2-pyridyldithio end group of the heterotelechelic PEG (CHO-Bz-PEG-SSpyl) thus prepared were characterized by (1)H and (13)C NMR, showing that the reaction proceeded almost quantitatively. The benzaldehyde end group is available to conjugate various ligands having a primary amino group by forming the pH-sensitive imine linkage (-N=CHC(6)H(4)-).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-(Diethoxymethyl)benzaldehyde, 97%