Merck
  • Review of fructosyl amino acid oxidase engineering research: a glimpse into the future of hemoglobin A1c biosensing.
Review of fructosyl amino acid oxidase engineering research: a glimpse into the future of hemoglobin A1c biosensing.
Journal of diabetes science and technology (2010-02-11)
Stefano Ferri, Seungsu Kim, Wakako Tsugawa, Koji Sode
RESUMO
Glycated proteins, particularly glycated hemoglobin A1c, are important markers for assessing the effectiveness of diabetes treatment. Convenient and reproducible assay systems based on the enzyme fructosyl amino acid oxidase (FAOD) have become attractive alternatives to conventional detection methods. We review the available FAOD-based assays for measurement of glycated proteins as well as the recent advances and future direction of FAOD research. Future research is expected to lead to the next generation of convenient, simple, and economical sensors for glycated protein, ideally suited for point-of-care treatment and self-monitoring applications.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Fructosyl-Amino Acid Oxidase from Corynebacterium sp., recombinant, expressed in E. coli, lyophilized powder, ≥0.45 units/mg protein