Merck
  • Interaction of MTHFR C677T and A1298C, and MTR A2756G gene polymorphisms in breast cancer risk in a population in Northeast Brazil.
Interaction of MTHFR C677T and A1298C, and MTR A2756G gene polymorphisms in breast cancer risk in a population in Northeast Brazil.
Anticancer research (2012-11-17)
Rita de Cássia Carvalho Barbosa, Débora Menezes da Costa, Denise Ellen Francelino Cordeiro, Ana Patricia Freitas Vieira, Silvia Helena Barem Rabenhorst
RESUMO
Polymorphisms in genes encoding enzymes of folate metabolism are a focus of breast cancer risk studies due of the role of these enzymes in DNA methylation, synthesis, and repair. MTHFR, encoding for 5,10-methylenetetrahydrofolate reductase, is one of the most studied genes in this regard, but findings are controversial, and the majority of studies have analyzed polymorphisms individually. In this case control study, we examined the combination of the polymorphisms MTHFR C677T and A1298C with MTR A2756G, where MTR, methionine synthase, is an important enzyme of the folate cycle in the methylation pathway. One hundred and forty-two patients with breast cancer and controls were included and the genotypes were determined using PCR-RFLP. In the population studied, individuals carrying the polymorphic allele in the heterozygous state for both enzymes, MTHFR C677T and MTR A2756G, had an increased risk [odds ratio, OR=2.77 (95% confidence interval, CI=1.19-6.52)] for disease, compared to those with the wild genotype. In addition, individuals carrying the MTR 2756 genotype AG had an increased risk when this was combined with the MTHFR 1298 genotype CC [OR=5.13 (95% CI=0.87-38.82)]. No significant results were found from the analyses associating the MTHFR C677T and A1298C genotypes. However, when stratifying the patients by age (50 years old as the cut-off), patients over 50 years old had greater risk, with the presence of both MTHFR polymorphisms in the heterozygous state [OR=5.33 (95% CI=1.42-21.03)]. This study points out the importance of the interactions between the MTHFR C677T, MTHFR A1298C and MTR A2756G polymorphisms, and also highlights the relevance of the MTR A2756G polymorphism and age in breast cancer risk.