Merck
Residue analysis of triflumizole and its metabolite in crops by high-performance liquid chromatography.
Journal of chromatography (1987-06-19)
N Shiga, O Matano, S Goto
RESUMO
A procedure for high-performance liquid chromatographic (HPLC) determination of triflumizole and its metabolite (Met) in crops was examined. Triflumizole and Met in sample crops were extracted with methanol and re-extracted into methylene chloride. After clean-up of the extract on a Florisil column, triflumizole and Met were determined by HPLC with UV detection at 238 nm. The HPLC column was packed with Nucleosil 5 C18 (ODS, 5 micron) and the eluent was acetonitrile-3 mM carbonate buffer (7:3, v/v, pH 9.0). The detection limit was 0.01-0.02 ppm and the recoveries from spiked crops (0.5 ppm) were 73-99% for triflumizole and 74-94% for Met. Triflumizole and Met were determined simultaneously and the method was shown to be applicable to residue analysis of these compounds in crops sprayed with Trifmine in fields.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Triflumizole, PESTANAL®, analytical standard