Merck
Autoimmune phenotype in patients with common variable immunodeficiency.
Journal of investigational allergology & clinical immunology (2013-11-23)
H Abolhassani, D Amirkashani, N Parvaneh, P Mohammadinejad, B Gharib, S Shahinpour, A Hirbod-Mobarakeh, M Mirghorbani, M Movahedi, M Gharagozlou, N Rezaei, A Aghamohammadi
RESUMO
Autoimmune disorders occur with a higher incidence in common variable immunodeficiency (CVID) patients than in the general population. To describe the clinical features of the autoimmune phenotype in patients with CVID. The hospital records of all diagnosed CVID patients referred to the Children's Medical Center Hospital in Tehran, Iran between 2000 and 2010 were reviewed. Patients were also classified according to the presence or absence of autoimmune disease. Of 52 patients studied, 26.9% (n=14) had shown at least 1 autoimmune manifestation during the study period. Autoimmune cytopenias and juvenile rheumatoid arthritis were the most common form of autoimmunity in our series. Autoimmunity was significantly associated with polyclonal lymphocytic infiltrative disorders (P = .017), increased serum Immunoglobulin (Ig) M levels (P < .001), decreased IgE values (P = .04) and diminished switched memory B-cell count (P < .001). Because autoimmunity is one of the first manifestations in CVID, humoral immune system tests should be considered in autoimmune patients with a history of recurrent infection. The presence of polyclonal lymphocytic infiltrative disorders and decreased switched memory B-cells may predispose CVID patients to autoimmunity.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Cotrimoxazole, Ready Made Solution, 100 mg/mL in DMSO