Merck
  • Efficacy and tolerability of fixed combination of brimonidine 0.2%/timolol 0.5% compared with fixed combination of dorzolamide 2%/timolol 0.5% in the treatment of patients with elevated intraocular pressure: a meta-analysis of randomized controlled trials.
Efficacy and tolerability of fixed combination of brimonidine 0.2%/timolol 0.5% compared with fixed combination of dorzolamide 2%/timolol 0.5% in the treatment of patients with elevated intraocular pressure: a meta-analysis of randomized controlled trials.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2013-02-06)
Patrick Budengeri, Jin-Wei Cheng, Ji-Ping Cai, Rui-Li Wei
RESUMO
To evaluate the efficacy and tolerability of the fixed-combination brimonidine 0.2%/timolol 0.5% (FCBT) compared with the fixed-combination dorzolamide 2%/timolol 0.5% (FCDT) in the treatment of patients with elevated intraocular pressure (IOP). Pertinent randomized controlled trials comparing FCBT with FCDT in patients with elevated IOP were identified through systematic searches of the Cochrane Library, PubMed, and EMBASE. The main efficacy measures were the IOP reduction (IOPR), including diurnal mean IOPR, and peak IOPR. The main tolerability measure was the individual adverse events. The pooled estimates and 95% confidence intervals (CIs) were carried out in RevMan version 5.0 software. Seven studies involving 582 patients were included in the meta-analysis. With a weighted mean difference (WMD) of IOPR in diurnal mean of 0.44 mmHg (95% CI, 0.00-0.88), the FCBT was as effective as FCDT in lowering IOP in patients with elevated IOP (P=0.05). The WMD of IOPR at peak was 0.65 mmHg (95% CI, -0.06 to 1.35) (P=0.76), and there was no significant difference between FCBT and FCDT. FCBT caused burning/stinging in less patients than FCDT [pooled relative risk: 0.45 (95% CI, 0.29-0.70)]. Both FCBT and FCDT can effectively lower IOP in patients with elevated IOP, and the IOP-lowering efficacy of FCBT is noninferior to that of FCDT. Additionally, FCBT affords an ocular comfort advantage compared with FCDT.
MATERIAIS
Número do produto
Marca
Descrição do produto
Brimonidine tartrate, European Pharmacopoeia (EP) Reference Standard